missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrofluoric Acid , Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 264808

 View more versions of this product

Catalog No. MK264810

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Hydrofluoric Acid
7664-39-3
FH
KRHYYFGTRYWZRS-UHFFFAOYSA-N
fluorane
14917
20.01
10lb.
52%
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
F
20.006
CHEBI:29228
Poly

Chemical Identifiers

7664-39-3
20.006
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
CHEBI:29228
F
FH
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
fluorane
Documents