Please login to your online account to display your discounted pricing

Hydrofluoric Acid , Macron Fine Chemicals™


macron

Quantity 1lb.
Packaging Circle of Safety Bottle

Manufacturer: avantor performance  264812

 View more versions of this product

Catalog No. MK26481

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 20.01
Quantity 1lb.
Molecular Formula HF
Chemical Name or Material Hydrofluoric Acid
Packaging Circle of Safety Bottle
Assay Percent Range 52%
CAS 7664-39-3

Specifications

Chemical Name or Material Hydrofluoric Acid
Assay Percent Range 52%
Molecular Formula HF
Formula Weight 20.01