Please login to your online account to display your discounted pricing

Hydrofluoric Acid , Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Hydrofluoric Acid
Assay Percent Range 52%
Molecular Formula HF
Formula Weight 20.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK26481

 
avantor performance
264812
Circle of Safety Bottle 1lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK264810

 
avantor performance
264808
Poly 10lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 20.01
Quantity 1lb.
Chemical Name or Material Hydrofluoric Acid
Assay Percent Range 52%
Molecular Formula HF
CAS 7664-39-3
Packaging Circle of Safety Bottle