Please login to your online account to display your discounted pricing

Hydrofluoric acid, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$430.33 - $845.80

Chemical Identifiers

Chemical Name or Material Hydrofluoric Acid
CAS 7664-39-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HF
Formula Weight 36.46
Synonym Fluoric Acid; Fluorhydric Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003071

 
avantor performance
956018
Each for $430.33

02003072

 
avantor performance
956006
Bagged Poly Bottle 4L Each for $845.80
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Assay 48.0 to 51.0%
Formula Weight 36.46
Chemical Name or Material Hydrofluoric acid
Specific Gravity 1L = 1.18kg
Melting Point −83.6°C
Boiling Point 19.5°C
Physical Form Liquid
Molecular Formula HF
CAS 7664-39-3
Synonym Fluoric acid; Fluorhydric acid
Density 1.15g/mL
Color Clear Colorless
Quantity 2.5L

    For Microelectronic Use