Please login to your online account to display your discounted pricing

Hydrofluoric Acid (Certified ACS), Fisher Chemical

$986.31 - $986.31

Chemical Identifiers

Chemical Name or Material Hydrofluoric Acid
CAS 7732-18-5
CAS 7664-39-3
Purity Grade Certified ACS Grade
Assay 48.0 to 51.0% (w/w)
Assay Percent Range 48.0 to 51.0 w/w %
Molecular Formula HF
Formula Weight 20.01
MDL Number MFCD00011346
Synonym Fluorhydric Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A147-10LB

  Accessories SDS
fisher scientific company
FLA14710LB
Poly Bottle 10 lb. Each for $986.31
Case of 4 Each for $3,945.24
Description & Specifications

Specifications

Chemical Name or Material Hydrofluoric Acid
Color Colorless
Quantity 10 lb.
pH 2.0
Cleanup Symbol Acid
CAS 7732-18-5
ChemAlert Storage Symbol White
Health Hazard 2 DANGER!
CAS 7664-39-3
Health Hazard 3 Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1
DOT Information DOT Class 8, : Corrosive
Arsenic (As) 0.05ppm max.
CAS 7732-18-5, 7664-39-3
Purity Grade Certified ACS Grade
Physical Form Liquid
Assay 48.0 to 51.0% (w/w)
Assay Percent Range 48.0 to 51.0 w/w %
Molecular Formula HF
Formula Weight 20.01
MDL Number MFCD00011346
Chloride 5ppm max.
UN Number 1790
Synonym Fluorhydric Acid
Density 2.21 (Air = 1.0)
Copper (Cu) 0.1ppm max.
Fluorosilicic Acid 0.01% max.
Heavy Metals (as Pb) 0.5ppm max.
Identification Pass Test
Iron (Fe) 1ppm max.
Lead (Pb) 0.1ppm max.
Nickel (Ni) 0.1ppm max.
Packaging Poly Bottle
Phosphate 1ppm max.
Residue after Ignition 5ppm max.
Sulfate and Sulfite 5ppm max.
Melting Point −35°C
Boiling Point 105°C
Safety and Handling
 

Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1

DANGER!