Learn More
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 423800250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Hydrofluoric acid | |
48.0-51.0% | |
7664-39-3,7732-18-5 | |
1.1500g/mL | |
Plastic bottle | |
1.15 | |
FH | |
HF | |
06,284; 09,238; 16,176 | |
Solubility in water: soluble. | |
F | |
20.01 | |
CHEBI:29228 | |
48.0 to 51.0% |
Colorless | |
-35.0°C | |
105.0°C | |
0.5ppm max. | |
0.0005% max. | |
25 g | |
Liquid | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
hydrogen fluoride | |
14917 | |
20 | |
ACS Reagent |
Chemical Identifiers
7664-39-3 | |
20.01 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
14917 | |
hydrogen fluoride |
FH | |
MFCD00011346 | |
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484 | |
CHEBI:29228 | |
F |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal if inhaled.
GHS P Statement:
Wear face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 231-634-8
RTECSNumber : MW7875000
TSCA : TSCA
RUO – Research Use Only