Please login to your online account to display your discounted pricing

Hydrofluoric acid, ACS, 48-51%, Alfa Aesar™

$80.85 - $80.85

Chemical Identifiers

Chemical Name or Material Hydrofluoric acid
Name Note Special handling precautions required
CAS 7664-39-3
Purity Grade ACS
Physical Form Liquid
Assay Percent Range 48-51%
Molecular Formula HF
Formula Weight 20.01g/mol
MDL Number MFCD00011346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3325836

 
alfa aesar
33258-36
500g Each for $80.85
Description & Specifications

Specifications

Appearance Colorless liquid
Density 1.16g/cm3
Odor Acrid
Color Colorless
Quantity 500g
Chemical Name or Material Hydrofluoric acid
Name Note Special handling precautions required
CAS 7664-39-3
Purity Grade ACS
Physical Form Liquid
Assay Percent Range 48-51%
Molecular Formula HF
Formula Weight 20.01g/mol
MDL Number MFCD00011346
Health Hazard 1 H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.
EINECS Number 231-634-8
Safety and Handling

H300-H310-H330-H314: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes severe skin burns and eye damage.