Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrofluoric acid, 99.99% (metals basis) 40% min

Chemical Identifiers

Chemical Name or Material Hydrofluoric acid
CAS 7664-39-3
Formula Weight 20.01
MDL Number MFCD00011346
UN Number UN1790
Merck Index 14,4790
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10989AC

  SDS
alfa aesar™
10989AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10989AK

  SDS
alfa aesar™
10989AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 20.01
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 25mL
Assay 99.99%
Color Colorless
MDL Number MFCD00011346
Health Hazard 1 Danger
Density 1.16g/mL
Merck Index 14,4790
TSCA TSCA
UN Number UN1790
Boiling Point 112°C
Assay Percent Notes (metals basis) 40% min
CAS 7664-39-3
Sensitivity Hygroscopic
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID
RTECS Number MW7875000
Odor Acrid
Health Hazard 2 GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Physical Form Liquid
Chemical Name or Material Hydrofluoric acid
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.