missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrofluoric acid, 99.99% (metals basis) 40% min

Manufacturer:  Alfa Aesar™ 10989AK

 View more versions of this product

Catalog No. AA10989AK



Specifications

Specifications

Hydrofluoric acid
250mL
(metals basis) 40% min
MFCD00011346
14,4790
KRHYYFGTRYWZRS-UHFFFAOYSA-N
fluorane
20.006
CHEBI:29228
Liquid
Acrid
112°C
Colorless
7664-39-3
FH
UN1790
antisal 2b
F
1.16g/mL
14917
20.01
99.99%
Hygroscopic

Chemical Identifiers

7664-39-3
20.006
KRHYYFGTRYWZRS-UHFFFAOYSA-N
14917
fluorane
FH
MFCD00011346
antisal 2b
CHEBI:29228
F
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID

RTECSNumber : MW7875000

TSCA : TSCA

Documents