Please login to your online account to display your discounted pricing

Hydrofluoric Acid, 48-50% (Technical), Fisher Chemical

$1,974.77 - $1,974.77

Chemical Identifiers

Chemical Name or Material Hydrofluoric Acid, 48-50%
CAS 7732-18-5, 7664-39-3
CAS 7664-39-3
Purity Grade Technical Grade
Assay 48 to 50% (w/w)
Assay Percent Range 48 to 50 w/w %
Molecular Formula HF
Formula Weight 20.01
MDL Number MFCD00011346
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A146-10LB

  Accessories SDS
fisher scientific company
FLA14610LB
Poly Bottle 10 lb. Case of 4 Each for $1,974.77
Description & Specifications

Specifications

Assay 48 to 50% (w/w)
Melting Point −35°C
Boiling Point 105°C
Purity Grade Technical Grade
Formula Weight 20.01
Quantity 10 lb.
DOT Information DOT Class 8, : Corrosive
CAS 7732-18-5, 7664-39-3
Density 2.21 (Air = 1.0)
Physical Form Liquid
Vapor Density 2.21g/mol
Color Colorless
pH 2.0
Health Hazard 2 DANGER!
CAS 7664-39-3
Health Hazard 3 Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1
Identification Pass Test
Chemical Name or Material Hydrofluoric Acid, 48-50%
Cleanup Symbol Acid
Assay Percent Range 48 to 50 w/w %
Molecular Formula HF
MDL Number MFCD00011346
ChemAlert Storage Symbol White
UN Number 1790
Packaging Poly Bottle
Safety and Handling
 

Emergency Overview
May be fatal if inhaled, absorbed through skin, or swallowed. Corrosive to metals. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:4
Flammability:0
Instability:1

DANGER!