Learn More
Hydrocinnamoyl chloride, 98%, Thermo Scientific Chemicals
$136.99 - $136.99
Chemical Identifiers
CAS | 645-45-4 |
---|---|
Molecular Formula | C9H9ClO |
Molecular Weight (g/mol) | 168.62 |
MDL Number | MFCD00000748 |
InChI Key | MFEILWXBDBCWKF-UHFFFAOYSA-N |
Synonym | 3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride |
PubChem CID | 64801 |
IUPAC Name | 3-phenylpropanoyl chloride |
SMILES | C1=CC=C(C=C1)CCC(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC134380250
|
Thermo Scientific Chemicals
134380250 |
25 g | Glass bottle |
Each for $136.99
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
645-45-4 | |
168.62 | |
MFEILWXBDBCWKF-UHFFFAOYSA-N | |
64801 | |
C1=CC=C(C=C1)CCC(=O)Cl |
C9H9ClO | |
MFCD00000748 | |
3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride | |
3-phenylpropanoyl chloride |
Specifications
645-45-4 | |
Colorless to Yellow | |
107°C (11.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5260 to 1.528 | |
MFCD00000748 | |
09, 511 | |
3-phenylpropionyl chloride, hydrocinnamoyl chloride, benzenepropanoyl chloride, dihydrocinnamoyl chloride, hydrocinnamyl chloride, propionyl chloride, 3-phenyl, hydrocinnamoylchloride, beta-phenylpropionyl chloride, 3-phenylpropionic acid chloride, beta-phenylpropanoyl chloride | |
MFEILWXBDBCWKF-UHFFFAOYSA-N | |
3-phenylpropanoyl chloride | |
64801 | |
98% | |
Hydrocinnamoyl chloride, 98% |
-6°C | |
1.1370g/mL | |
108°C | |
97.5% min. (GC) | |
C9H9ClO | |
C6H5(CH2)2COCl | |
25 g | |
1.137 | |
Solubility in water: decomposes in water. Other solubilities: soluble in ether | |
C1=CC=C(C=C1)CCC(=O)Cl | |
168.62 | |
168.62 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-443-6
RTECSNumber : MW5652395
TSCA : TSCA
RUO – Research Use Only