Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrochlorothiazide, 98%

$372.50 - $372.50

Chemical Identifiers

Chemical Name or Material Hydrochlorothiazide
CAS 58-93-5
Molecular Formula C7H8ClN3O4S2
Formula Weight 297.74
MDL Number MFCD00051765
Beilstein 625101
Merck Index 14,4781
Synonym 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2209322

 
alfa aesar™
B2209322
100g Each for $372.50
Description & Specifications

Specifications

Formula Weight 297.74
Chemical Name or Material Hydrochlorothiazide
TSCA TSCA
Assay 98%
Odor Odorless
Molecular Formula C7H8ClN3O4S2
MDL Number MFCD00051765
CAS 58-93-5
Beilstein 625101
Merck Index 14,4781
Synonym 6-Chloro-7-sulfamyl-3,4-dihydro-1,2,4-benzothiadiazine-1,1-dioxide
RTECS Number DK9100000
Melting Point 266° to 272°C
Quantity 100g