missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, TraceGrade, 34-37%, Spectrum™ Chemical

H1122, 7647-01-0, HCl

$317.32 - $1752.97

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603212
View Documents Promotion Details
Spectrum Chemical Mfg Cor
H1122500ML
500 mL Amber Glass Bottle
Each for $317.32
 
18603214
View Documents Promotion Details
Spectrum Chemical Mfg Cor
H112225LTPL
2.5 L Poly Bottle
Each for $507.97
 
18603213
View Documents Promotion Details
Spectrum Chemical Mfg Cor
H1122500MLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $1,470.32
 
18603215
View Documents Promotion Details
Spectrum Chemical Mfg Cor
H112225LTPLCS4
4 x 2.5 L Poly Bottle
Case of 4 Each for $1,752.97
 
Description

Description

Spectrum™ Chemical Hydrochloric Acid, TraceGrade is a strong, highly corrosive mineral acid with extremely low metal content (over 50 with levels less than 1 PPB). This grade is primarily used as an analytical reagent in elemental analysis determinations using ICP and AA techniques, digestion of water samples and as a doping agent for organic semiconductors.
Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
ClH
MFCD00011324 MFCD00792839
hydrogen chloride

Specifications

7647-01-0,7732-18-5
Amber Glass Bottle
ClH
UN1789
Cl
36.46
High Purity Solvent
70%,30%
500 mL
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
34 to 37%
SDS
missing translation for 'documents'

missing translation for 'documents'