Please login to your online account to display your discounted pricing

Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical


Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Quantity 1L
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSA50-1

 View more versions of this product

Catalog No. SA50-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 1L
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Clear
pH 0.10
Concentration or Composition (by Analyte or Components) 0.5N (N/2)
UN Number 1789
CAS 7732-18-5
Molecular Formula HCl
Concentration 0.498 to 0.502N
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
ChemAlert Storage Symbol White
MDL Number MFCD00011324
Cleanup Symbol Acid
Traceability to NIST LOT 84L Potassium acid phthalate
Packaging Poly Bottle
Synonym Muriatic Acid
Identification Pass Test
Health Hazard 2 CAUTION!
Health Hazard 1 Signal Word:CAUTION!
CAS 7647-01-0
Chemical Name or Material Hydrochloric Acid Solution, 0.5N (N/2)

Specifications

Chemical Name or Material Hydrochloric Acid Solution, 0.5N (N/2)
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
Molecular Formula HCl
MDL Number MFCD00011324
Synonym Muriatic Acid
Accessories

Accessories for Catalog Number: SA501

Item Mfr. No. Item Description Price Per Unit Quantity  
17-253
Uvex™ Classic™ Chemical-Splash Goggles
north safety products
UVS360
Integrated ventilation system Each for $6.49
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

CAUTION!

 

SDS & Safety Documents