missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Hydrochloric acid, pure, fuming, 37% solution in water, Thermo Scientific™

Hydrochloric acid, 35.0 to 39.0%, ClH, CAS Number-7647-01-0, 7732-18-5, acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 57.0 deg.C, CHEBI:17883

Supplier:  Thermo Scientific Chemicals 124620250

Catalog No. AC124620250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Hydrochloric acid
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
36.46
CHEBI:17883
Solution
Pure
5ppm max.
Plastic drum
1.18
-35.0°C
Undesignated
1.9 mPa.s (15°C)
7647-01-0,7732-18-5
HCl
04,450; 05,533; 06,283
Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
[H+].[Cl-]
1.1800g/mL
313
36.46
35.0 to 39.0%
10ppm max. (Cl)
2ppm max.
Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
100ppm max.
57.0°C
25L
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Product Certifications
Promotions

Promotions

RUO – Research Use Only