Please login to your online account to display your discounted pricing

Hydrochloric acid, pure, 25% solution in water, ACROS Organics™


Quantity 2.5L

Manufacturer: thermo fisher scientific  AC387790025

 View more versions of this product

Catalog No. AC387790025

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Health Hazard 2 GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage.
Assay ≥25.0%
Physical Form Liquid
Formula Weight 36.45
Quantity 2.5L
Color Clear colorless
Purity Grade Pure
Density 1.1200g/mL
Residue after Ignition 0.002% max.
Linear Formula HCl
Health Hazard 1 Danger
MDL Number MFCD00011324
Chemical Name or Material Hydrochloric acid
Sulfate 0.002% max.
Iron (Fe) 0.0001% max.
Heavy Metals (as Pb) 1ppm max.
CAS 7732-18-5
Synonym Muriatic acid, 25%
CAS 7647-01-0
Molecular Formula ClH

Specifications

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Pure
Assay ≥25.0%
Molecular Formula ClH
Linear Formula HCl
Formula Weight 36.45
MDL Number MFCD00011324
Synonym Muriatic acid, 25%
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage.

SDS & Safety Documents