Please login to your online account to display your discounted pricing

Hydrochloric Acid NF/FCC/ACS, Macron Fine Chemicals™


macron

Quantity 4 x 2.5L
Packaging Glass

Manufacturer: avantor performance  261244

 View more versions of this product

Catalog No. MK2612212X4

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
Quantity 4 x 2.5L
Name Note (HCl 36.5%-38.0%)
CAS 7647-01-0
Purity Grade ACS/FCC/NF
Molecular Formula HCl
Formula Weight 36.46
Packaging Glass

Specifications

Chemical Name or Material Hydrochloric Acid
Name Note (HCl 36.5%-38.0%)
CAS 7647-01-0
Purity Grade ACS/FCC/NF
Molecular Formula HCl
Formula Weight 36.46