Please login to your online account to display your discounted pricing

Hydrochloric Acid, Multi-Compendial, N.F., J.T.Baker™

$172.01 - $907.59

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Purity Grade Multi-Compendial/NF
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003066

 
avantor performance
954403
Glass Bottle 2.5L Each for $907.59

02003067

 
avantor performance
954405
Glass Bottle, Single ship 2.5L Each for $227.51

02003068

 
avantor performance
954433
pvc coated glass bottle 2.5L Each for $298.33

02003069

 
avantor performance
954402
Glass Bottle 500mL Each for $172.01
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Quantity 2.5L
CAS 7647-01-0
Purity Grade Multi-Compendial/NF
Molecular Formula HCl
Formula Weight 36.46
Packaging Glass Bottle
Specific Gravity 1L = 1.18kg