Please login to your online account to display your discounted pricing

Hydrochloric Acid, with Methyl Orange, Certified, 33% (v/v) ±1%, LabChem™

$191.37 - $191.37

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Max % 86.3195
CAS 7647-01-0
CAS Max % 13.68
CAS 547-58-0
CAS Max % 0.0005
Name Note with Methyl Orange
Purity Grade Certified Grade
Linear Formula HCl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC151204

 
lab chem
LC151204
4L Each for $191.37
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 33% ± 1% (v/v)
Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
CAS Max % 0.0005
Solubility Soluble in water
Specific Gravity 1.07g/mL
Viscosity 1.16cSt
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Name Note with Methyl Orange
Physical Form Liquid
Linear Formula HCl
CAS 7732-18-5
UN Number UN1789
Density 1.07g/mL
Health Hazard 1 Danger
CAS Max % 86.3195
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
CAS Max % 13.68
CAS 547-58-0
Packaging Poly Bottle
Color Clear, pink
Quantity 4L
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.