missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, J.T.Baker™

$797.79 - $797.79

Chemical Identifiers

CAS 7647-01-0
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
Synonym Muriatic acid solution
Chemical Name or Material Hydrochloric acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
14-650-319 Avantor Performance
690005
500mL Each for $797.79
14650386 Avantor Performance
H61314
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
14650387 Avantor Performance
H61346
2.5L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MKH61316 Avantor Performance
H61316
2.5L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7647-01-0
HCl
Muriatic acid solution
Liquid
36.46
Hydrochloric acid

Specifications

Clear colorless
7647-01-0
HCl
Muriatic acid solution
500mL
Liquid
36.46
Hydrochloric acid