Please login to your online account to display your discounted pricing

Hydrochloric Acid, in Isopropanol, Certified, 0.200N ±0.002N (0.2M), LabChem™

$60.17 - $518.49

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 67-63-0
CAS Max % 99.28
CAS 7647-01-0
CAS Max % 0.72
Name Note in Isopropanol
Purity Grade Certified Grade
Linear Formula HCl
Formula Weight 36.46
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC154102

 
lab chem
LC154102
1L Each for $60.17

LC154104

 
lab chem
LC154104
4L Each for $193.00

LC154105

 
lab chem
LC154105
20L Each for $518.49
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Color Clear, colorless
Quantity 1L
Name Note in Isopropanol
CAS 67-63-0
Health Hazard 1 Danger
CAS Max % 99.28
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 0.72
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula HCl
Formula Weight 36.46
UN Number UN2924
Concentration 0.200N ±0.002N (0.2M)
Density 785kg/m3
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Specific Gravity 785kg/m3
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Flash Point 12°C
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Melting Point −88°C
Boiling Point 82°C
Safety and Handling

Danger

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.