Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrochloric acid, Environmental Grade Plus, 33-36%

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0
Purity Grade Environmental Grade Plus
Formula Weight 36.46
MDL Number MFCD00011324
UN Number UN1789
Merck Index 14,4780
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38744AP

  SDS
alfa aesar™
38744AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid, Double Sub-Boiling Quartz Distillation
Formula Weight 36.46
Chemical Name or Material Hydrochloric acid
TSCA TSCA
Boiling Point 81.5° to 110°C
Purity Grade Environmental Grade Plus
Assay 33 to 36%
MDL Number MFCD00011324
CAS 7647-01-0
UN Number UN1789
Merck Index 14,4780
Density 1.19g/mL
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROCHLORIC ACID
Odor Pungent
RTECS Number MW4025000
Quantity 500mL
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H335
Causes severe skin burns and eye damage.
May cause respiratory irritation.