Please login to your online account to display your discounted pricing

Hydrochloric acid, Environmental Grade, 34-37.5%, Alfa Aesar™

single sub-boiling quartz distillation

$113.62 - $198.89

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
Name Note Single sub-boiling quartz distillation
CAS 7647-01-0
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range 34-37.5%
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38743K4

  SDS
alfa aesar
38743K4
2.5L Each for $198.89

AA38743AP

  SDS
alfa aesar
38743AP
500mL Each for $113.62
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric acid
Color Clear colorless
Quantity 2.5L
Name Note Single sub-boiling quartz distillation
CAS 7647-01-0
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Appearance Colorless liquid
DOT Information Hazard Class: 8, Packing Group: II
EINECS Number 231-595-7
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range 34-37.5%
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
Odor Pungent
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).