Please login to your online account to display your discounted pricing

Hydrochloric Acid, EMD Millipore

$180.77 - $180.77

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note OmniPure™
Linear Formula HCl in water
Formula Weight 36.46
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

36-103-1083

 
emd millipore corporation
HX06071DEL
Each for $180.77
Description & Specifications

Specifications

Melting Point −74°C
Boiling Point 110°C
Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
Name Note OmniPure™
Assay Percent Range 34 to 37%
Linear Formula HCl in water
CAS 7647-01-0
Synonym Muriatic Acid
Recommended Storage Room Temperature
Packaging Poly Coated Glass Bottle
Safety and Handling

Room Temperature