missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid Concentrate, (Certified) ACS, Fisher Chemical

Each bottle contains exact amount of 1N concentrate required to make 1 liter of 0.1N (N/10) solution

Manufacturer:  Fisher Chemical FLSA55

Catalog No. SA55




Description

Description

Specifications

Specifications

Hydrochloric Acid Concentrate, 1N
100mL
7647-01-0,7732-18-5
MFCD00011324
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
[H+].[Cl-]
hydrogen chloride
36.46
CHEBI:17883
Certified
Glass Bottle
-17°C
Undesignated
1.1
ClH
1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
0.0990 to 0.1010N
1.16g/mL
313
Liquid
Pass Test
LOT 84L Potassium acid phthalate
81.5°C to 110°C

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
[H+].[Cl-]
Safety and Handling

Safety and Handling

DOTInformation : Class 8: Corrosive

SDS Documents
Product Certifications