Learn More
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Supplier: Fisher Chemical A144612GAL
Specifications
1 | |
-35°C | |
57°C | |
0.1ppm max. | |
Glass Carboy | |
1.18 | |
6.5 gal. | |
Liquid | |
Soluble in water | |
Cl | |
36.46 |
Colorless | |
7647-01-0 | |
1.18g/cm³ | |
Pass Test | |
4ppm max. | |
125mbar at 20°C | |
ClH | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
36.5 to 38.0% (w/w) |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrogen chloride |
Safety and Handling
DANGER!
Emergency Overview
Causes burns by all exposure routes. Use only under a chemical fume hood. Wear personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Do not induce vomiting. Do not induce vomiting. Call a physician or Poison Control Center immediately.
NFPA
Health:3
Flammability:0
Instability:1