Please login to your online account to display your discounted pricing

Hydrochloric Acid, Certified, 1.0% (v/v) ±0.1%, LabChem™

$28.77 - $94.83

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Max % 99.56
CAS 7647-01-0
CAS Max % 0.44
Purity Grade Certified Grade
Linear Formula HCl
Formula Weight 36.46
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC150001

 
lab chem
LC150001
500mL Each for $28.77

LC150002

 
lab chem
LC150002
1L Each for $45.97

LC150004

 
lab chem
LC150004
4L Each for $94.83
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Color Clear, colorless
Quantity 500mL
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 99.56
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS Max % 0.44
Purity Grade Certified Grade
Physical Form Liquid
Assay 1.0% ± 0.1% (v/v)
Linear Formula HCl
Formula Weight 36.46
Synonym Muriatic Acid
Density 1g/mL
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen chloride
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.