Please login to your online account to display your discounted pricing

Hydrochloric Acid, Certified, 0.5165N ±0.0030N (0.5165M), LabChem™

$70.77 - $235.60

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Max % 98.13
CAS 7647-01-0
CAS Max % 1.87
Purity Grade Certified Grade
Linear Formula HCl
Formula Weight 36.46
UN Number UN1789
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC152904

 
lab chem
LC152904
4L Each for $70.77

LC152905

 
lab chem
LC152905
20L Each for $235.60
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric Acid
Color Clear, colorless
Quantity 4L
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 98.13
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 1.87
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula HCl
Formula Weight 36.46
UN Number UN1789
Synonym Muriatic Acid
Concentration 0.5165N ±0.0030N (0.5165M)
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.