Please login to your online account to display your discounted pricing

Hydrochloric Acid, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

$30.31 - $234.94

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Max % 99.82
CAS 7647-01-0
CAS Max % 0.18
Purity Grade Certified Grade
Linear Formula HCl
Formula Weight 36.46
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC152004

 
lab chem
LC152004
4L Each for $69.46

LC152005

 
lab chem
LC152005
20L Each for $234.94

LC152002

 
lab chem
LC152002
1L Each for $30.31
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Decomposition Information Hydrogen chloride
Physical Form Liquid
Linear Formula HCl
CAS 7732-18-5
Synonym Muriatic Acid
Concentration 0.0500N ±0.0005N (0.05M)
Density 1g/mL
Health Hazard 1 Warning
CAS Max % 99.82
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Identification Passes Test
CAS Max % 0.18
Packaging Poly Bottle
Color Clear, colorless
Quantity 4L
Safety and Handling

Warning

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.