Please login to your online account to display your discounted pricing

Hydrochloric Acid, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$122.30 - $1,521.14

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003042

 
avantor performance
938505
Narrow Mouth PTFE Bottle 2.5L Each for $240.07

02003054

 
avantor performance
938503
Pressure Bottom Poly Bottle 4L Each for $1,521.14

02003057

 
avantor performance
938501
Poly Bottle 500mL Each for $122.30
Description & Specifications

Specifications

Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Molecular Formula HCl
Specific Gravity 1L = 1.18kg
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Packaging Narrow Mouth PTFE Bottle
Quantity 2.5L

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use