Please login to your online account to display your discounted pricing

Hydrochloric acid, AR Select™ Plus, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
Synonym Muriatic acid solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

14650246

 
avantor performance
V07804
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

14650512

 
avantor performance
V07846
SAFEMOR Container 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Hydrochloric acid
Color Clear colorless
Quantity 500mL
CAS 7647-01-0
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
Synonym Muriatic acid solution