missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, for analysis, fuming, 37% solution in water, Thermo Scientific Chemicals

$97.35 - $186.40

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124635000
SDS Accessories View Documents Promotion Details
Thermo Scientific Chemicals
124635000
500 mL Glass Bottle
Each for $97.35
Case of 6 Each for $584.10
 
AC124630010
SDS Accessories View Documents Promotion Details
Thermo Scientific Chemicals
124630010
1 L Glass Bottle
Each for $136.32
Case of 4 Each for $545.28
 
AC124630026
SDS View Documents Promotion Details
Thermo Scientific Chemicals
124630026
2.5 L Plastic bottle
Each for $186.40
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
Cl
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883

Specifications

-35.0°C
57.0°C
2ppm max.
(SO2) Passes Test
500 mL
ClH
HCl
04,450; 05,533; 06,283
Solubility in water: miscible: 823g/L (0°C); 561g/L (60°C). Other solubilities: soluble in alcohol, benzene and ether, insoluble in hydrocarbons
Cl
36.46
CHEBI:17883
35.0 to 39.0%
Hydrochloric acid
7647-01-0,7732-18-5
1.1800g/mL
Glass Bottle
1.18
1.9 mPa.s (15°C)
Solution
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
Analytical
Safety and Handling

Safety and Handling

GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May be corrosive to metals.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Absorb spillage to prevent material damage. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only