Learn More
Hydrochloric acid, for analysis, 25% solution in water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 387800025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hydrochloric acid | |
-70.0°C | |
107.0°C | |
Plastic bottle | |
1.12 | |
ClH | |
HCl | |
15, 4818 | |
Solubility in water: soluble. | |
Cl | |
36.46 | |
CHEBI:17883 | |
≥25.0% |
Colorless | |
7647-01-0,7732-18-5 | |
1.1200g/mL | |
0.0005% max. | |
2.5 L | |
Liquid | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.45 | |
Analytical |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Safety and Handling
GHS H Statement
May cause respiratory irritation. Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
RUO – Research Use Only