missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, ACS Grade, 36.5 to 38.0%, LabChem™

Manufacturer:  LabChem LC149503

 View more versions of this product

Catalog No. LC149503



Specifications

Specifications

Hydrochloric Acid
2.5L
63,37
HCl
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
1190kg/m3
chlorane
313
36.46
36.5 to 38.0%
Passes Test
Soluble in water
Room Temperature
Colorless
7647-01-0,7732-18-5
ClH
1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Cl
36.458
CHEBI:17883
Liquid
ACS
Clear Glass Bottle
1190kg/m3
Hydrogen chloride

Chemical Identifiers

7647-01-0
36.458
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, spray, vapors.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Us

Danger

DOTInformation : Class 8

EINECSNumber : 231-595-7

Documents