Please login to your online account to display your discounted pricing

Hydrochloric acid, Acculute Std. Volumetric Soln., Final Concentration 1.0N, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Hydrochloric acid
Physical Form Liquid
Molecular Formula HCl
Formula Weight 36.46
MDL Number MFCD00011324
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA88559LQ

  SDS
alfa aesar
88559LQ
6 units CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA88559LS

  SDS
alfa aesar
88559LS
6 units CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Formula Weight 36.46
Quantity 6 units
DOT Information Hazard Class: 8, Packing Group: II
Chemical Name or Material Hydrochloric acid
EINECS Number 231-595-7
Appearance Colorless liquid
Molecular Formula HCl
MDL Number MFCD00011324
Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Odor Acidic
Safety and Handling

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).