Learn More
Hydrochloric acid, 99.999% (metals basis), 36.5% min, Thermo Scientific Chemicals
$187.49 - $671.64
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | chlorane |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA87617AK
|
Thermo Scientific Chemicals
087617AK |
250 mL |
Each for $187.49
|
|
|||||
AA87617AU
|
Thermo Scientific Chemicals
087617AU |
1000 mL |
Each for $455.12
|
|
|||||
AA87617K4
|
Thermo Scientific Chemicals
87617K4 |
2.5 L |
Each for $671.64
|
|
|||||
Description
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
Colorless liquid | |
7647-01-0 | |
1.19 g/mL | |
99.999% (metals basis) | |
ClH | |
MFCD00011324 MFCD00792839 | |
Completely miscible with water. | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid |
Colorless | |
81.5°C to 110°C | |
Pungent | |
250 mL | |
Liquid | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8, Packing Group: II
EINECSNumber : 231-595-7
RTECSNumber : MW4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only