missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Hydrochloric Acid, 99.99% Trace Metals Basis, Honeywell™

37 wt. % in H2O, 99.999% trace metals basis

$422.26 - $422.26

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.458
MDL Number MFCD00011324
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name chlorane
SMILES Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60046312 View Documents Honeywell™
339253500ML
500mL Poly Bottle Each for $422.26
 
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications

Chemical Identifiers

7647-01-0
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

Colorless
>0.1 (at 20°C)
ClH
UN1789
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
1.2g/mL at 25°C
313
36.46g/mol
Stinging
3.23psi (21.1°C), 7.93psi (37.7°C)
Liquid
42°C
500mL
7647-01-0
MFCD00011324
1098214
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
36.458
CHEBI:17883
99.999% trace metals basis
Poly Bottle
1.3 (vs air), 1.3 (vs air)
-35°C
Hydrochloric acid
Safety and Handling

Safety and Handling

P261-P280-P305 + P351 + P338-P310

DOTInformation : Class 8/ PGII

RTECSNumber : 231-595-7