Please login to your online account to display your discounted pricing

Hydrochloric Acid, 75% (v/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 24.0
CAS Max % 26.0
CAS 7647-01-0
CAS Min % 74.0
CAS Max % 76.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3580-75-1

  SDS
ricca chemical company
RI3580.751
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3580-75-16

 
ricca chemical company
RI3580.7516
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

3580-75-32

 
ricca chemical company
RI3580.7532
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight greenish yellow
Quantity 4L
Chemical Name or Material Hydrochloric Acid
Concentration or Composition Notes 75% (v/v)
CAS 7732-18-5
CAS Min % 74.0
Solvent or Matrix Aqueous Solution
CAS Min % 24.0
CAS Max % 26.0
CAS 7647-01-0
CAS Max % 76.0
Packaging Poly natural