Please login to your online account to display your discounted pricing

Hydrochloric Acid, 6N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$122.05 - $270.95

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note 6N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003063

 
avantor performance
561902
Narrow Mouth Polyethylene Bottle 1L Each for $122.05

02003064

 
avantor performance
561903
Pressure Bottom Poly Bottle 4L Each for $270.95
Description & Specifications

Specifications

Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
Molecular Formula HCl
Specific Gravity 1L = 1.10kg
Name Note 6N Volumetric Solution
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L
Purity Grade BAKER ANALYZED™ Reagent
CAS 7647-01-0