missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric acid, 6.0N Standardized Solution, Alfa Aesar™

Manufacturer:  Alfa Aesar 44921K2

 View more versions of this product

Catalog No. AA44921K2



Specifications

Specifications

Hydrochloric acid
Colorless to pale yellow liquid
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46
23 hPa (17mm Hg) at 20°C
1L
7647-01-0,7732-18-5
ClH
Hydrochloric acid: 49.6%; Water: 50.4%
Cl
36.458
CHEBI:17883
Liquid
Colorless to Yellow

Chemical Identifiers

7647-01-0
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
CHEBI:17883
Cl
ClH
MFCD00011324
313
chlorane
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

DOTInformation : Hazard Class: 8, Packing Group: II

EINECSNumber : 231-595-7

SDS Documents