Please login to your online account to display your discounted pricing

Hydrochloric Acid 6.0N Solution, Macron Fine Chemicals™

macron

$247.60 - $247.60

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid 6.0N Solution
CAS 7647-01-0
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MKH1681

 
avantor performance
H16805
Glass Bottle 1L Each for $247.60
Case of 6 Each for $1,485.60

MKH1684

 
avantor performance
H16807
Glass Bottle 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Hydrochloric Acid 6.0N Solution
CAS 7647-01-0
Packaging Glass Bottle