Please login to your online account to display your discounted pricing

Hydrochloric Acid, 6.0N Solution, Biotech Reagent, J.T.Baker™

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

$246.55 - $1,389.22

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0
Name Note 6.0N Solution
Purity Grade Biotech Reagent Grade
Molecular Formula HCl
Formula Weight 36.46
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003061

 
avantor performance
032707
Poly Bottle 19L Each for $1,389.22

02003062

 
avantor performance
032702
Narrow Mouth Polyethylene Bottle 1L Each for $246.55
Description & Specifications

Specifications

Purity Grade Biotech Reagent Grade
Formula Weight 36.46
Chemical Name or Material Hydrochloric Acid
Molecular Formula HCl
CAS 7647-01-0
Specific Gravity 1L = 1.10kg
Name Note 6.0N Solution
Packaging Poly Bottle
Quantity 19L