missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrochloric Acid, 50% (v/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI35805PT

 View more versions of this product

Catalog No. 35805PT



Description

Description

Specifications

Specifications

Hydrochloric Acid
Colorless to Green-Yellow
7647-01-0,7732-18-5
51.0,51.0
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
Cl
36.458
CHEBI:17883
Glass
50% (v/v)
2.50L
49.0,49.0
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
Liquid
Aqueous Solution

Chemical Identifiers

7647-01-0
36.458
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt
CHEBI:17883
Cl
ClH
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
SDS
Documents
Accessories