missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 50% (v/v) Aqueous Solution, Ricca Chemical
Manufacturer: Ricca Chemical Company RI358016
Specifications
Hydrochloric Acid | |
Colorless to Green-Yellow | |
7647-01-0,7732-18-5 | |
51.0,51.0 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
Cl | |
36.458 | |
CHEBI:17883 | |
Poly natural |
50% (v/v) | |
500mL | |
49.0,49.0 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
Liquid | |
Aqueous Solution |
Chemical Identifiers
7647-01-0 | |
36.458 | |
acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |