Learn More
Hydrochloric acid, 5.0N Standardized Solution, Thermo Scientific Chemicals
$82.23 - $433.01
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Description
Hydrochloric acid, 5.0N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
CHEBI:17883 |
ClH | |
MFCD00011324 MFCD00792839 | |
313 | |
Cl |
Specifications
7647-01-0,7732-18-5 | |
Colorless to Yellow | |
MFCD00011324 MFCD00792839 | |
Cl | |
36.46 | |
CHEBI:17883 | |
Hydrochloric acid: 41.3%; Water: 58.7% | |
Miscible with water, methanol,ethanol and ether | |
Hydrochloric acid |
1 L | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 231-595-7
RTECSNumber : MW4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only