missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 5.00 Normal, Ricca Chemical
Manufacturer: Ricca Chemical Company RIR37400001A
Specifications
Hydrochloric Acid | |
1L | |
66.0,32.21 | |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
Liquid | |
5.00Normal |
Colorless to Green-Yellow | |
7647-01-0,7732-18-5 | |
68.0,33.53 | |
acide chlorhydrique | |
Cl | |
36.458 | |
CHEBI:17883 | |
Poly Natural |
Chemical Identifiers
7647-01-0 | |
36.458 | |
acide chlorhydrique | |
CHEBI:17883 | |
Cl |
ClH | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
chlorane |
Safety and Handling
ShelfLife : 18 months