Please login to your online account to display your discounted pricing

Hydrochloric acid, 37%, extra pure

$56.63 - $56.63

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0, 7732-18-5
CAS 7732-18-5
Purity Grade Extra Pure
Molecular Formula ClH
Formula Weight 36.45
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AC450560050

 
thermo fisher scientific
AC450560050
Each for $56.63
Description & Specifications

Specifications

Melting Point −35.0°C
Formula Weight 36.45
Quantity 5L
Chemical Name or Material Hydrochloric Acid
Boiling Point 57.0°C
Purity Grade Extra Pure
CAS 7647-01-0, 7732-18-5
Synonym Muriatic Acid
Density 1.1800g/mL
Packaging Plastic Drum
Assay Percent Range 37%
Molecular Formula ClH
CAS 7732-18-5