Please login to your online account to display your discounted pricing

Hydrochloric acid, 37%, for analysis, max. 0.000001% Hg

$84.36 - $84.36

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0, 7732-18-5
CAS 7732-18-5
Purity Grade Extra Pure
Assay Percent Range 37%
Molecular Formula ClH
Formula Weight 36.45
Synonym Muriatic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC450550025

  SDS
thermo fisher scientific
AC450550025
Glass Bottle 2.5L Each for $84.36
Description & Specifications

Specifications

Formula Weight 36.45
Melting Point −35.0°C
Quantity 2.5L
Chemical Name or Material Hydrochloric Acid
CAS 7647-01-0, 7732-18-5
Synonym Muriatic Acid
Packaging Glass Bottle
Purity Grade Extra Pure
Boiling Point 57.0°C
Density 1.1800g/mL
Assay Percent Range 37%
Molecular Formula ClH
CAS 7732-18-5