missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hydrochloric Acid, 3.85 N, Ricca Chemical
Supplier: Ricca Chemical Company R37280001A
Specifications
Hydrochloric Acid | |
71.0,26.56 | |
Natural Poly Bottle | |
ClH | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
[H+].[Cl-] | |
36.46 | |
CHEBI:17883 |
7647-01-0,7732-18-5 | |
74.0,27.64 | |
1 L | |
MFCD00011324 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 |
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
[H+].[Cl-] |