Please login to your online account to display your discounted pricing

Hydrochloric Acid, 3.85 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
CAS Min % 71.0
CAS Max % 74.0
CAS 7647-01-0
CAS Min % 26.56
CAS Max % 27.64
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R37280001A

 
ricca chemical company
RIR37280001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R37280004A

 
ricca chemical company
RIR37280004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slight greenish yellow
Quantity 1L
CAS Min % 71.0
CAS Max % 27.64
Chemical Name or Material Hydrochloric Acid
CAS 7732-18-5
Concentration or Composition Notes 3.85Normal
CAS Max % 74.0
CAS Min % 26.56
Packaging Poly natural
CAS 7647-01-0